Methyl 2,3-difluoro-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

A1232995-100MG A1232995-1G A1232995-50MG A1232995-250MG
76679-804EA 231.01 USD
76679-804 76679-806 76679-808 77592-400
Methyl 2,3-difluoro-4-nitrobenzoate 97%
Methyl 2,3-difluoro-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅F₂NO₄
MDL Number: MFCD28790655
CAS Number: 1803786-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR