(2-(4-(Bromomethyl)phenyl)ethene-1,1,2-triyl)tribenzene 98%

Supplier: Ambeed
Danger

A1232668-1G A1232668-250MG A1232668-25G A1232668-5G
76679-792EA 66 USD
76679-792 76679-794 76679-796 76679-798
(2-(4-(Bromomethyl)phenyl)ethene-1,1,2-triyl)tribenzene 98%
(2-(4-(Bromomethyl)phenyl)ethene-1,1,2-triyl)tribenzene

Precaución: for Research Use Only

Formula: C₂₇H₂₁Br
MDL Number: MFCD31619167
CAS Number: 1361969-01-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR