1-Phenyl-1H-imidazole trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-1H-imidazol-3-ium trifluoromethanesulphonate

A1232551-1G A1232551-25G A1232551-5G
76679-786EA 15 USD
76679-786 76679-788 76679-790
1-Phenyl-1H-imidazole trifluoromethanesulfonate 98%
1-Phenyl-1H-imidazole trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃N₂O₃S
MDL Number: MFCD32206610
CAS Number: 361447-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR