2,4,6-Tris(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,5-triazine 98%
Supplier: Ambeed
|
Warning
|
A1232281-100MG
A1232281-1G
A1232281-250MG
A1232281-5G
76679-776EA
14
USD
76679-776
76679-778
76679-780
76679-782
2,4,6-Tris(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,5-triazine 98%
2,4,6-Tris(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,5-triazine
Precaución: for Research Use Only
Formula:
C₃₉H₄₈B₃N₃O₆ |
MDL Number:
MFCD32670809 CAS Number: 1447947-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...