Bis(morpholinothiocarbonyl) disulfide 98%

Supplier: Ambeed
Danger

A1230676-10MG A1230676-1MG A1230676-25MG A1230676-50MG A1230676-5MG
76679-752EA 53 USD
76679-752 76679-754 76679-756 76679-758 76679-760
Bis(morpholinothiocarbonyl) disulfide 98%
Bis(morpholinothiocarbonyl) disulfide

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂S₄
MDL Number: MFCD00072237
CAS Number: 729-46-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR