1-Benzyl 4-(tert-butyl) (S)-2-methylpiperazine-1,4-dicarboxylate 98%

Supplier: Ambeed
Warning

A1230549-1G A1230549-250MG A1230549-25G A1230549-5G
76679-744EA 77.45 USD
76679-744 76679-746 76679-748 76679-750
1-Benzyl 4-(tert-butyl) (S)-2-methylpiperazine-1,4-dicarboxylate 98%
1-Benzyl 4-(tert-butyl) (S)-2-methylpiperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₄
MW: 334.4101 g/mol
MDL Number: MFCD03840835
CAS Number: 1163793-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR