2-(tert-Butyl(2-((di-tert-butylphosphino)methyl)benzyl)phosphino)pyridine 97%
Supplier: Ambeed
|
Warning
|
A1229869-100MG
A1229869-250MG
A1229869-25MG
A1229869-50MG
76679-728EA
349
USD
76679-728
76679-730
76679-732
76679-734
2-(tert-Butyl(2-((di-tert-butylphosphino)methyl)benzyl)phosphino)pyridine 97%
2-(tert-Butyl(2-((di-tert-butylphosphino)methyl)benzyl)phosphino)pyridine
Precaución: for Research Use Only
Formula:
C₂₅H₃₉NP₂ MW: 415.5314 g/mol |
CAS Number:
2410085-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...