(2R,4R)-Pentane-2,4-diylbis(bis(3,5-dimethylphenyl)phosphine) 97%

Supplier: Ambeed
Warning

A1229125-100MG A1229125-1G A1229125-250MG
76679-710EA 111.4 USD
76679-710 76679-712 76679-714
(2R,4R)-Pentane-2,4-diylbis(bis(3,5-dimethylphenyl)phosphine) 97%
(2R,4R)-Pentane-2,4-diylbis(bis(3,5-dimethylphenyl)phosphine)

Precaución: for Research Use Only

Formula: C₃₇H₄₆P₂
MW: 552.7087 g/mol
MDL Number: MFCD28100578
CAS Number: 217648-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR