3-[[(Benzylthio)carbonothioyl]thio]propionic acid 98%

Supplier: Ambeed
Warning

A1228894-100MG A1228894-250MG A1228894-1G A1228894-25G
76679-706EA 30 USD
76679-706 76679-708 77592-392 77866-884
3-[[(Benzylthio)carbonothioyl]thio]propionic acid 98%
3-[[(Benzylthio)carbonothioyl]thio]propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂S₃
MDL Number: MFCD30475390
CAS Number: 497931-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR