2-(((9H-Purin-6-yl)amino)methyl)phenol 97%
Supplier: Ambeed
|
Warning
|
A1228689-100MG
A1228689-1G
A1228689-250MG
76679-690EA
109.28
USD
76679-690
76679-692
76679-694
2-(((9H-Purin-6-yl)amino)methyl)phenol 97%
2-(((9H-Purin-6-yl)amino)methyl)phenol
Precaución: for Research Use Only
Formula:
C₁₂H₁₁N₅O MW: 241.2486 g/mol |
MDL Number:
MFCD18641954 CAS Number: 20366-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...