Pyrimidine-2-thiol 98%

Supplier: Ambeed
Warning

A1228679-100G A1228679-25G A1228679-500G A1228679-5G
76679-682EA 206 USD
76679-682 76679-684 76679-686 76679-688
Pyrimidine-2-thiol 98%
Pyrimidine-2-thiol

Precaución: for Research Use Only

Formula: C₄H₄N₂S
MDL Number: MFCD00006073
CAS Number: 131242-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR