1,2-BIs((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A1228632-100MG A1228632-1G A1228632-250MG
76679-670EA 143.23 USD
76679-670 76679-672 76679-674
1,2-BIs((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%
1,2-BIs((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂₃H₃₈F₃O₃P₂RhS
MW: 616.4593 g/mol
CAS Number: 213343-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR