(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,5R)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane
A1228474-100MG
A1228474-1G
A1228474-250MG
A1228474-5G
76679-654EA
68.96
USD
76679-654
76679-656
76679-658
76679-660
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane ≥97%, ee 99%
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Precaución: for Research Use Only
Formula:
C₁₅H₂₁O₂P MW: 264.305 g/mol |
MDL Number:
MFCD09842713 CAS Number: 1044256-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...