N-((5,6-Dichloro-1H-benzo[d]imidazol-2-yl)methyl)-9-(1-methyl-1H-pyrazol-4-yl)-2-morpholino-9H-purin-6-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
SR-4835
A1228338-10MG
A1228338-25MG
A1228338-50MG
A1228338-100MG
76679-644EA
54.11
USD
76679-644
76679-646
76679-648
77571-390
N-((5,6-Dichloro-1H-benzo[d]imidazol-2-yl)methyl)-9-(1-methyl-1H-pyrazol-4-yl)-2-morpholino-9H-purin-6-amine 98%
N-((5,6-Dichloro-1H-benzo[d]imidazol-2-yl)methyl)-9-(1-methyl-1H-pyrazol-4-yl)-2-morpholino-9H-purin-6-amine
Precaución: for Research Use Only
Formula:
C₂₁H₂₀Cl₂N₁₀O MW: 499.36 g/mol |
MDL Number:
MFCD32263441 CAS Number: 2387704-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...