4-(2-(1H-Imidazol-1-yl)ethoxy)benzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A1228336-10MG A1228336-1MG A1228336-25MG A1228336-50MG A1228336-5MG
76679-634EA 42 USD
76679-634 76679-636 76679-638 76679-640 76679-642
4-(2-(1H-Imidazol-1-yl)ethoxy)benzoic acid hydrochloride 97%
4-(2-(1H-Imidazol-1-yl)ethoxy)benzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₂O₃
MDL Number: MFCD00475663
CAS Number: 74226-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR