2-([1,1'-Biphenyl]-4-yl)-4-(4-bromophenyl)-6-phenyl-1,3,5-triazine 97%
Supplier: Ambeed
|
Warning
|
A1228284-1G
A1228284-5G
76679-630EA
27.59
USD
76679-630
76679-632
2-([1,1'-Biphenyl]-4-yl)-4-(4-bromophenyl)-6-phenyl-1,3,5-triazine 97%
2-([1,1'-Biphenyl]-4-yl)-4-(4-bromophenyl)-6-phenyl-1,3,5-triazine
Precaución: for Research Use Only
Formula:
C₂₇H₁₈BrN₃ MW: 464.3559 g/mol |
CAS Number:
1911641-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...