tert-Butyl (4-oxocycloheptyl)carbamate 97%
Supplier: Ambeed
|
Danger
|
A1228000-250MG
A1228000-100MG
76679-622EA
178
USD
76679-622
77866-873
tert-Butyl (4-oxocycloheptyl)carbamate 97%
tert-Butyl (4-oxocycloheptyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₃ |
MDL Number:
MFCD24470690 CAS Number: 1031335-15-5 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...