2-Chloro-4,6-diiodophenol 97%

Supplier: Ambeed
Warning

A1227771-1G A1227771-250MG
76679-610EA 39 USD
76679-610 76679-612
2-Chloro-4,6-diiodophenol 97%
2-Chloro-4,6-diiodophenol

Precaución: for Research Use Only

Formula: C₆H₃ClI₂O
MDL Number: MFCD00672958
CAS Number: 15459-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR