1-Bromo-3-chloro-4-iodo-2-methylbenzene 97%
Supplier: Ambeed
|
Warning
|
A1227338-1G
A1227338-250MG
A1227338-5G
76679-604EA
141.1
USD
76679-604
76679-606
76679-608
1-Bromo-3-chloro-4-iodo-2-methylbenzene 97%
1-Bromo-3-chloro-4-iodo-2-methylbenzene
Precaución: for Research Use Only
Formula:
C₇H₅BrClI MW: 331.376 g/mol |
MDL Number:
MFCD30478263 CAS Number: 1799612-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...