3-(5-Amino-1H-pyrazol-3-yl)benzonitrile 97%

Supplier: Ambeed
Warning

A1227320-100MG A1227320-1G A1227320-250MG A1227320-25G A1227320-5G
76679-586EA 25.47 USD
76679-586 76679-588 76679-590 76679-592 77621-156
3-(5-Amino-1H-pyrazol-3-yl)benzonitrile 97%
3-(5-Amino-1H-pyrazol-3-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈N₄
MW: 184.1973 g/mol
MDL Number: MFCD32856365
CAS Number: 1800082-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR