4-Bromo-1,6-dimethyl-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one 97%

Supplier: Ambeed
Warning

A1227310-100MG A1227310-1G
76679-582EA 299.18 USD
76679-582 76679-584
4-Bromo-1,6-dimethyl-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one 97%
4-Bromo-1,6-dimethyl-1,6-dihydro-7H-pyrrolo[2,3-c]pyridin-7-one

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
MW: 241.0846 g/mol
CAS Number: 2412985-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR