Benzene-1,2,4,5-tetracarboxamide 98%

Supplier: Ambeed
Warning

A1227200-1G A1227200-25G
76679-578EA 21.22 USD
76679-578 76679-580
Benzene-1,2,4,5-tetracarboxamide 98%
Benzene-1,2,4,5-tetracarboxamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₄O₄
MW: 250.2108 g/mol
MDL Number: MFCD00450905
CAS Number: 6183-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR