Ethyl 2,4-dichlorophenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(2,4-dichlorophenyl)ethanoate, Ethyl (2,4-dichlorophenyl)acetate, Ethyl 2-(2,4-dichlorophenyl)acetate

A1227070-100MG A1227070-1G A1227070-250MG A1227070-5G
76679-568EA 5.31 USD
76679-568 76679-570 76679-572 76679-574
Ethyl 2,4-dichlorophenylacetate 98%
Ethyl 2,4-dichlorophenylacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
MDL Number: MFCD01566538
CAS Number: 41022-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR