4''-Ethyl-2',3,4,5-tetrafluoro-1,1':4',1''-terphenyl 97%

Supplier: Ambeed
Warning

A1227062-1G A1227062-250MG A1227062-5G
76679-562EA 39.26 USD
76679-562 76679-564 76679-566
4''-Ethyl-2',3,4,5-tetrafluoro-1,1':4',1''-terphenyl 97%
4''-Ethyl-2',3,4,5-tetrafluoro-1,1':4',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₀H₁₄F₄
MW: 330.3188 g/mol
CAS Number: 326894-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR