(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3'-chloro-[1,1'-biphenyl]-4-yl)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
A1226968-100MG
A1226968-250MG
76679-554EA
85.94
USD
76679-554
76679-556
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3'-chloro-[1,1'-biphenyl]-4-yl)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3'-chloro-[1,1'-biphenyl]-4-yl)propanoic acid
Precaución: for Research Use Only
Formula:
C₃₀H₂₄ClNO₄ MW: 497.9689 g/mol |
MDL Number:
MFCD29917524 CAS Number: 1380437-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...