6-Methyl-4-(methylthio)-2-oxo-1,2-dihydropyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A1226962-100MG A1226962-1G A1226962-250MG A1226962-5G
76679-546EA 27.59 USD
76679-546 76679-548 76679-550 76679-552
6-Methyl-4-(methylthio)-2-oxo-1,2-dihydropyridine-3-carbonitrile 97%
6-Methyl-4-(methylthio)-2-oxo-1,2-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂OS
MW: 180.2269 g/mol
MDL Number: MFCD16656995
CAS Number: 57663-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR