4,4'-(1,2-Diphenylethene-1,2-diyl)dianiline 98%

Supplier: Ambeed
Warning

A1226924-100MG A1226924-1G A1226924-250MG
76679-540EA 12 USD
76679-540 76679-542 76679-544
4,4'-(1,2-Diphenylethene-1,2-diyl)dianiline 98%
4,4'-(1,2-Diphenylethene-1,2-diyl)dianiline

Precaución: for Research Use Only

Formula: C₂₆H₂₂N₂
CAS Number: 99094-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR