RHC-80267 98%

Supplier: Ambeed
Warning

Synonyms: Cyclohexanone O-((6-((((cyclohexylideneamino)oxy)carbonyl)amino)hexyl)carbamoyl) oxime, RHC 80267

A1226769-10MG A1226769-1MG A1226769-25MG A1226769-50MG
76679-528EA 36 USD
76679-528 76679-530 76679-532 76679-534
RHC-80267 98%
RHC-80267

Precaución: for Research Use Only

Formula: C₂₀H₃₄N₄O₄
MW: 394.516 g/mol
MDL Number: MFCD00210844
CAS Number: 83654-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR