Methyl 6-methyl-3-(methylamino)picolinate 97%

Supplier: Ambeed
Warning

A1226743-1G A1226743-250MG
76679-524EA 144 USD
76679-524 76679-526
Methyl 6-methyl-3-(methylamino)picolinate 97%
Methyl 6-methyl-3-(methylamino)picolinate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.2038 g/mol
MDL Number: MFCD18259079
CAS Number: 1256837-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR