3-(5-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1226590-100MG A1226590-1G A1226590-250MG A1226590-5G A1226590-25G
76679-510EA 6 USD
76679-510 76679-512 76679-514 76679-516 77571-376
3-(5-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%
3-(5-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrN₂O₃
MDL Number: MFCD32263142
CAS Number: 1010100-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR