5',5'''-Bis(4-carboxyphenyl)-5''-(4,4''-dicarboxy-[1,1':3',1''-terphenyl]-5'-yl)-[1,1':3',1'':3'',1''':3''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A1226469-100MG A1226469-1G A1226469-250MG A1226469-5G
76679-496EA 44.56 USD
76679-496 76679-498 76679-500 76679-502
5',5'''-Bis(4-carboxyphenyl)-5''-(4,4''-dicarboxy-[1,1':3',1''-terphenyl]-5'-yl)-[1,1':3',1'':3'',1''':3''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid 98%
5',5'''-Bis(4-carboxyphenyl)-5''-(4,4''-dicarboxy-[1,1':3',1''-terphenyl]-5'-yl)-[1,1':3',1'':3'',1''':3''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₆₆H₄₂O₁₂
MW: 1027.0325 g/mol
CAS Number: 1331745-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR