5-(4-(Diphenylamino)phenyl)thiophene-2-carbaldehyde 98%

Supplier: Ambeed
Warning

A1226418-100MG A1226418-250MG A1226418-5G A1226418-1G
76679-490EA 13 USD
76679-490 76679-492 76679-494 77621-150
5-(4-(Diphenylamino)phenyl)thiophene-2-carbaldehyde 98%
5-(4-(Diphenylamino)phenyl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₂₃H₁₇NOS
MDL Number: MFCD32667030
CAS Number: 291279-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR