3',4',5',6'-Tetrakis(4-formylphenyl)-[1,1':2',1''-terphenyl]-4,4''-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1226417-100MG A1226417-250MG A1226417-5G A1226417-1G
76679-484EA 33.95 USD
76679-484 76679-486 76679-488 77566-318
3',4',5',6'-Tetrakis(4-formylphenyl)-[1,1':2',1''-terphenyl]-4,4''-dicarbaldehyde 98%
3',4',5',6'-Tetrakis(4-formylphenyl)-[1,1':2',1''-terphenyl]-4,4''-dicarbaldehyde

Precaución: For research use only.

Formula: C₄₈H₃₀O₆
MW: 702.748 g/mol
MDL Number: MFCD32667051
CAS Number: 1862220-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR