[1,1':3',1''-Terphenyl]-4,4''-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1226416-250MG A1226416-5G A1226416-1G
76679-480EA 69 USD
76679-480 76679-482 77866-854
[1,1':3',1''-Terphenyl]-4,4''-dicarbaldehyde 98%
[1,1':3',1''-Terphenyl]-4,4''-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₂
MDL Number: MFCD00182938
CAS Number: 171820-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR