4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzaldehyde 98%

Supplier: Ambeed
Warning

A1226415-100MG A1226415-1G A1226415-250MG A1226415-5G
76679-474EA 26.53 USD
76679-474 76679-476 76679-478 77621-148
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzaldehyde 98%
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzaldehyde

Precaución: for Research Use Only

Formula: C₃₃H₁₈O₃
MW: 462.4942 g/mol
MDL Number: MFCD32667055
CAS Number: 2289758-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR