4',4''',4'''''-(1,3,5-Triazine-2,4,6-triyl)tris(([1,1'-biphenyl]-4-carbaldehyde)) 98%

Supplier: Ambeed
Warning

A1226414-1G A1226414-250MG A1226414-100MG A1226414-5G
76679-470EA 82 USD
76679-470 76679-472 77866-852 77866-853
4',4''',4'''''-(1,3,5-Triazine-2,4,6-triyl)tris(([1,1'-biphenyl]-4-carbaldehyde)) 98%
4',4''',4'''''-(1,3,5-Triazine-2,4,6-triyl)tris(([1,1'-biphenyl]-4-carbaldehyde))

Precaución: for Research Use Only

Formula: C₄₂H₂₇N₃O₃
MDL Number: MFCD32667034
CAS Number: 1221509-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR