4,4'-(1,4-Phenylenebis(ethyne-2,1-diyl))dibenzaldehyde 98%

Supplier: Ambeed
Warning

A1226413-100MG A1226413-1G A1226413-250MG
76679-464EA 32.89 USD
76679-464 76679-466 76679-468
4,4'-(1,4-Phenylenebis(ethyne-2,1-diyl))dibenzaldehyde 98%
4,4'-(1,4-Phenylenebis(ethyne-2,1-diyl))dibenzaldehyde

Precaución: for Research Use Only

Formula: C₂₄H₁₄O₂
MW: 334.3668 g/mol
MDL Number: MFCD32667010
CAS Number: 192188-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR