2',5'-Dibutoxy-[1,1':4',1''-terphenyl]-4,4''-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1226412-100MG A1226412-1G A1226412-250MG
76679-458EA 19 USD
76679-458 76679-460 76679-462
2',5'-Dibutoxy-[1,1':4',1''-terphenyl]-4,4''-dicarbaldehyde 98%
2',5'-Dibutoxy-[1,1':4',1''-terphenyl]-4,4''-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₂₈H₃₀O₄
MDL Number: MFCD32667018
CAS Number: 1501954-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR