2',5'-Bis(hexyloxy)-[1,1':4',1''-terphenyl]-4,4''-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1226410-100MG A1226410-1G A1226410-250MG
76679-446EA 22 USD
76679-446 76679-448 76679-450
2',5'-Bis(hexyloxy)-[1,1':4',1''-terphenyl]-4,4''-dicarbaldehyde 98%
2',5'-Bis(hexyloxy)-[1,1':4',1''-terphenyl]-4,4''-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₃₂H₃₈O₄
MW: 486.6400 g/mol
MDL Number: MFCD32667029
CAS Number: 474974-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR