4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayltetrakis(ethyne-2,1-diyl))tetraaniline 97%

Supplier: Ambeed
Warning

A1226409-100MG A1226409-250MG A1226409-1G
76679-442EA 36.08 USD
76679-442 76679-444 77566-316
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayltetrakis(ethyne-2,1-diyl))tetraaniline 97%
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayltetrakis(ethyne-2,1-diyl))tetraaniline

Precaución: For research use only.

Formula: C₄₈H₃₀N₄
MW: 662.7786 g/mol
MDL Number: MFCD32667025
CAS Number: 1404196-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR