4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))trianiline 98%

Supplier: Ambeed
Warning

A1226407-1G A1226407-250MG A1226407-5G
76679-430EA 160.2 USD
76679-430 76679-432 76679-434
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))trianiline 98%
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))trianiline

Precaución: for Research Use Only

Formula: C₃₀H₂₁N₃
MW: 423.5079 g/mol
MDL Number: MFCD32667002
CAS Number: 326002-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR