[1,1':4',1'':4'',1'''-Quaterphenyl]-3,3''',5,5'''-tetracarboxylic acid 98%

Supplier: Ambeed
Warning

A1226403-100MG A1226403-1G A1226403-250MG
76679-420EA 31.83 USD
76679-420 76679-422 76679-424
[1,1':4',1'':4'',1'''-Quaterphenyl]-3,3''',5,5'''-tetracarboxylic acid 98%
[1,1':4',1'':4'',1'''-Quaterphenyl]-3,3''',5,5'''-tetracarboxylic acid

Precaución: for Research Use Only

Formula: C₂₈H₁₈O₈
MW: 482.4377 g/mol
MDL Number: MFCD32667041
CAS Number: 921619-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR