1,3,6,8-Tetrakis((trimethylsilyl)ethynyl)pyrene 98%
Supplier: Ambeed
|
Warning
|
A1226401-100MG
A1226401-1G
A1226401-250MG
A1226401-5G
76679-412EA
32.89
USD
76679-412
76679-414
76679-416
76679-418
1,3,6,8-Tetrakis((trimethylsilyl)ethynyl)pyrene 98%
1,3,6,8-Tetrakis((trimethylsilyl)ethynyl)pyrene
Precaución: for Research Use Only
Formula:
C₃₆H₄₂Si₄ MW: 587.0607 g/mol |
MDL Number:
MFCD21606941 CAS Number: 870258-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...