2,4,6-Tris(4-ethynylphenyl)-1,3,5-triazine 98%

Supplier: Ambeed
Warning

A1226400-250MG A1226400-5G A1226400-100MG A1226400-1G
76679-408EA 33 USD
76679-408 76679-410 77621-144 77621-146
2,4,6-Tris(4-ethynylphenyl)-1,3,5-triazine 98%
2,4,6-Tris(4-ethynylphenyl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₇H₁₅N₃
MDL Number: MFCD32667033
CAS Number: 425629-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR