4'',5''-Bis(4'-carboxy[1,1'-biphenyl]-4-yl)[1,1':4',1'':2'',1''':4''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A1226395-100MG A1226395-250MG
76679-398EA 23 USD
76679-398 76679-400
4'',5''-Bis(4'-carboxy[1,1'-biphenyl]-4-yl)[1,1':4',1'':2'',1''':4''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid 98%

Precaución: for Research Use Only

Formula: C₅₈H₃₈O₈
MDL Number: MFCD32667028
CAS Number: 1643112-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR