5'''-(4''-Carboxy-[1,1':4',1''-terphenyl]-4-yl)-[1,1':4',1'':4'',1''':3''',1'''':4'''',1''''':4''''',1''''''-septiphenyl]-4,4''''''-dicarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A1226392-100MG
A1226392-1G
A1226392-250MG
76679-390EA
44.56
USD
76679-390
76679-392
76679-394
5'''-(4''-Carboxy-[1,1':4',1''-terphenyl]-4-yl)-[1,1':4',1'':4'',1''':3''',1'''':4'''',1''''':4''''',1''''''-septiphenyl]-4,4''''''-dicarboxylic acid 98%
5'''-(4''-Carboxy-[1,1':4',1''-terphenyl]-4-yl)-[1,1':4',1'':4'',1''':3''',1'''':4'''',1''''':4''''',1''''''-septiphenyl]-4,4''''''-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₆₃H₄₂O₆ MW: 895.0040 g/mol |
CAS Number:
1246015-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...