6-Chloro-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazine 96%
Supplier: Ambeed
|
Warning
|
A1226379-100MG
A1226379-250MG
A1226379-5G
A1226379-1G
76679-384EA
84
USD
76679-384
76679-386
76679-388
77866-848
6-Chloro-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazine 96%
6-Chloro-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazine
Precaución: for Research Use Only
Formula:
C₁₃H₇ClF₃N₃O |
MDL Number:
MFCD22200828 CAS Number: 928337-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...