5-(3,5-Di-tert-butyl-4-hydroxybenzyl)thiazolidine-2,4-dione 98%

Supplier: Ambeed
Warning

Synonyms: NL-1

A1226353-10MG A1226353-1MG A1226353-50MG
76679-368EA 45.62 USD
76679-368 76679-370 76679-372
5-(3,5-Di-tert-butyl-4-hydroxybenzyl)thiazolidine-2,4-dione 98%
5-(3,5-Di-tert-butyl-4-hydroxybenzyl)thiazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₃S
MW: 335.46 g/mol
MDL Number: MFCD32671359
CAS Number: 188532-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR