1-(2,6-Dibromo-4-isopropylphenyl)-3-(pyridin-3-yl)urea 98%

Supplier: Ambeed
Warning

Synonyms: BX430

A1226351-10MG A1226351-1MG A1226351-25MG A1226351-50MG A1226351-5MG
76679-358EA 40.32 USD
76679-358 76679-360 76679-362 76679-364 76679-366
1-(2,6-Dibromo-4-isopropylphenyl)-3-(pyridin-3-yl)urea 98%
1-(2,6-Dibromo-4-isopropylphenyl)-3-(pyridin-3-yl)urea

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br₂N₃O
MW: 413.1071 g/mol
MDL Number: MFCD02678819
CAS Number: 688309-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR