5-(tert-Butyl)benzo[b]thiophene 98%

Supplier: Ambeed
Warning

A1226323-1G A1226323-25G A1226323-5G
76679-352EA 14.86 USD
76679-352 76679-354 76679-356
5-(tert-Butyl)benzo[b]thiophene 98%
5-(tert-Butyl)benzo[b]thiophene

Precaución: for Research Use Only

Formula: C₁₂H₁₄S
MW: 190.3046 g/mol
MDL Number: MFCD18451788
CAS Number: 17515-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR